Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIANP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244014.1 | NP_001230943.1 | 3 | 642 | ||
NM_001244015.1 | NP_001230944.1 | 3 | 708 | |||
NM_153685.3 | NP_710152.1 | 3 | 642 | |||
XM_011520926.2 | XP_011519228.1 | 3 | 599 | |||
GenBank mRNA | AK056295.1 | - | 3 | 599 | ||
AK091596.1 | - | 4 | 364 | |||
AK091656.1 | - | 3 | 442 | |||
AK095854.1 | - | 3 | 708 | |||
AK289648.1 | - | 3 | 497 | |||
AY358534.1 | - | 2 | 426 | |||
BC035736.1 | - | 3 | 445 | 5730150 | ||
DB496995.1 | - | 4 | 408 |