Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 6 | 515 | ||
XM_006721162.1 | XP_006721225.1 | 6 | 526 | |||
XM_011522928.1 | XP_011521230.1 | 6 | 693 | |||
XM_011522929.1 | XP_011521231.1 | 6 | 558 | |||
XM_011522930.2 | XP_011521232.1 | 6 | 526 | |||
XM_011522931.2 | XP_011521233.1 | 6 | 526 | |||
XM_017023012.1 | XP_016878501.1 | 6 | 542 | |||
XM_017023013.1 | XP_016878502.1 | 6 | 689 | |||
XM_017023014.1 | XP_016878503.1 | 3 | 223 | |||
GenBank mRNA | AJ441184.1 | - | 6 | 514 | ||
AK056456.1 | - | 6 | 514 | |||
AK074168.1 | - | 6 | 513 | |||
AK128387.1 | - | 6 | 611 | |||
AK293794.1 | - | 5 | 465 | |||
AK315590.1 | - | 6 | 518 | |||
BC013735.1 | - | 4 | 301 | |||
BC032542.1 | - | 6 | 521 | 5534376 |