Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 2 | 78 | ||
XM_006721162.1 | XP_006721225.1 | 2 | 89 | |||
XM_011522928.1 | XP_011521230.1 | 2 | 256 | |||
XM_011522929.1 | XP_011521231.1 | 2 | 121 | |||
XM_011522930.2 | XP_011521232.1 | 2 | 89 | |||
XM_011522931.2 | XP_011521233.1 | 2 | 89 | |||
XM_017023012.1 | XP_016878501.1 | 2 | 105 | |||
XM_017023013.1 | XP_016878502.1 | 2 | 252 | |||
GenBank mRNA | AJ441184.1 | - | 2 | 78 | ||
AK056456.1 | - | 2 | 78 | |||
AK074168.1 | - | 2 | 76 | |||
AK128387.1 | - | 2 | 78 | |||
AK293794.1 | - | 2 | 79 | |||
AK315590.1 | - | 2 | 80 | |||
BC032542.1 | - | 2 | 84 | 5534376 |