Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145080.3 | NP_659547.2 | 7 | 759 | ||
XM_006721023.3 | XP_006721086.1 | 8 | 913 | |||
XM_011545761.2 | XP_011544063.1 | 7 | 777 | |||
XM_011545762.1 | XP_011544064.1 | 9 | 1034 | |||
XM_017023040.1 | XP_016878529.1 | 9 | 1480 | |||
XM_017023041.1 | XP_016878530.1 | 10 | 1166 | |||
XM_017023042.1 | XP_016878531.1 | 8 | 1257 | |||
GenBank mRNA | AF161451.1 | - | 7 | 716 | ||
AF161455.1 | - | 7 | 729 | |||
AK056795.1 | - | 9 | 1480 | |||
AK315674.1 | - | 7 | 703 | |||
BC018938.2 | - | 6 | 645 | |||
BU685363.1 | - | 2 | 317 |