Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001967.3 | NP_001958.2 | 8 | 858 | ||
GenBank mRNA | AB209021.1 | - | 3 | 2547 | ||
AF208852.1 | - | 7 | 696 | |||
AK130087.1 | - | 8 | 853 | |||
AK225646.1 | - | 8 | 853 | |||
AK296279.1 | - | 4 | 392 | |||
AL117412.1 | - | 7 | 718 | |||
AL137681.1 | - | 3 | 1423 | |||
AL556981.3 | - | 7 | 718 | |||
BC012547.1 | - | 8 | 860 | 4079371 | ||
BC013708.1 | - | 8 | 838 | 4341420 | ||
BC015842.1 | - | 8 | 857 | 4337581 | ||
BC016295.2 | - | 2 | 136 | |||
BC048105.1 | - | 8 | 856 | 4820774 | ||
BT009860.1 | - | 8 | 819 | |||
D30655.1 | - | 8 | 834 | |||
S79942.1 | - | 8 | 834 |