Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030666.3 | NP_109591.1 | 7 | 1346 | ||
NR_073111.1 | 8 | 1478 | ||||
NR_073112.1 | 7 | 1342 | ||||
XM_011514333.1 | XP_011512635.1 | 8 | 1569 | |||
XM_011514334.2 | XP_011512636.1 | 7 | 1480 | |||
XM_011514335.2 | XP_011512637.1 | 7 | 1335 | |||
GenBank mRNA | AK291327.1 | - | 7 | 1159 | ||
AK298044.1 | - | 8 | 1288 | |||
AK304641.1 | - | 7 | 1191 | |||
AK312973.1 | - | 7 | 1157 | |||
BC009015.1 | - | 7 | 1144 | 3456154 | ||
BM785367.1 | - | 1 | 125 | |||
BT006928.1 | - | 6 | 1100 | |||
EU794624.1 | - | 6 | 1100 | |||
GQ472232.1 | - | 7 | 1144 | |||
M93056.1 | - | 7 | 1148 |