Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145353.1 | NP_001138825.1 | 8 | 1525 | ||
NM_172373.3 | NP_758961.1 | 9 | 1742 | |||
XM_005266276.1 | XP_005266333.1 | 9 | 1631 | |||
XM_005266277.3 | XP_005266334.1 | 9 | 2174 | |||
XM_011534950.1 | XP_011533252.1 | 11 | 1980 | |||
XM_011534951.2 | XP_011533253.1 | 10 | 2293 | |||
XM_017020409.1 | XP_016875898.1 | 11 | 1960 | |||
XM_017020410.1 | XP_016875899.1 | 10 | 1897 | |||
XM_017020411.1 | XP_016875900.1 | 10 | 1843 | |||
XM_017020412.1 | XP_016875901.1 | 10 | 1971 | |||
XM_017020413.1 | XP_016875902.1 | 10 | 2025 | |||
GenBank mRNA | AK095466.1 | - | 1 | 54 | ||
AK304289.1 | - | 8 | 1468 | |||
BC030507.2 | - | 8 | 1597 | 4385989 | ||
BX640798.1 | - | 6 | 1023 | |||
M82882.1 | - | 9 | 1684 |