Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005502.3 | NP_005493.2 | 48 | 6785 | ||
XM_005251773.2 | XP_005251830.1 | 48 | 6700 | |||
XM_005251776.2 | XP_005251833.1 | 47 | 6540 | |||
XM_011518339.2 | XP_011516641.1 | 49 | 6775 | |||
XM_011518340.2 | XP_011516642.1 | 50 | 6990 | |||
XM_011518341.2 | XP_011516643.1 | 49 | 6769 | |||
XM_011518342.2 | XP_011516644.1 | 46 | 6478 | |||
XM_017014378.1 | XP_016869867.1 | 49 | 6973 | |||
XM_017014379.1 | XP_016869868.1 | 49 | 7381 | |||
XM_017014380.1 | XP_016869869.1 | 49 | 6707 | |||
XM_017014381.1 | XP_016869870.1 | 49 | 6773 | |||
XM_017014382.1 | XP_016869871.1 | 47 | 7238 | |||
GenBank mRNA | AB055982.1 | - | 47 | 6381 | ||
AB208839.1 | - | 10 | 2763 | |||
AB445477.1 | - | 47 | 6381 | |||
AF165281.1 | - | 46 | 6321 | |||
AF285167.1 | - | 48 | 6671 | |||
AJ012376.1 | - | 46 | 6321 | |||
AK027864.1 | - | 13 | 1508 | |||
BC146856.1 | - | 48 | 6595 | 9056784 |