Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127197.1 | NP_001120669.1 | 4 | 594 | ||
NM_001421.3 | NP_001412.1 | 4 | 676 | |||
XM_005262389.3 | XP_005262446.1 | 4 | 1012 | |||
XM_011531307.2 | XP_011529609.1 | 3 | 422 | |||
XM_011531308.2 | XP_011529610.1 | 2 | 204 | |||
GenBank mRNA | AK314972.1 | - | 4 | 593 | ||
BC017194.2 | - | 4 | 559 | 3138355 | ||
CN314421.1 | - | 4 | 591 | |||
U32645.1 | - | 4 | 673 |