Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128210.1 | NP_001121682.1 | 6 | 1396 | ||
NM_181784.2 | NP_861449.2 | 6 | 1749 | |||
XM_005264201.1 | XP_005264258.1 | 5 | 1164 | |||
XM_017003576.1 | XP_016859065.1 | 4 | 1315 | |||
GenBank mRNA | AK025550.1 | - | 2 | 753 | ||
AK056479.1 | - | 6 | 1313 | |||
AK299079.1 | - | 6 | 1249 | |||
AK307714.1 | - | 4 | 895 | |||
AY299090.1 | - | 6 | 1410 | |||
BC029300.1 | - | 6 | 1326 | |||
BC111495.1 | - | 6 | 1737 | |||
BC130292.1 | - | 6 | 1346 | 40146323 | ||
BC136334.1 | - | 6 | 1346 | 9020321 |