Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128210.1 | NP_001121682.1 | 4, 5 | 630 | ||
NM_181784.2 | NP_861449.2 | 4, 5 | 983 | |||
XM_005264200.4 | XP_005264257.2 | 4, 5 | 1060 | |||
XM_005264201.1 | XP_005264258.1 | 3, 4 | 398 | |||
XM_005264202.4 | XP_005264259.1 | 4, 5 | 1060 | |||
XM_017003576.1 | XP_016859065.1 | 2, 3 | 549 | |||
GenBank mRNA | AK056479.1 | - | 4, 5 | 547 | ||
AK299079.1 | - | 4, 5 | 483 | |||
AY299090.1 | - | 4, 5 | 644 | |||
BC029300.1 | - | 4, 5 | 560 | |||
BC111495.1 | - | 4, 5 | 971 | |||
BC130292.1 | - | 4, 5 | 580 | 40146323 | ||
BC136334.1 | - | 4, 5 | 580 | 9020321 |