Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174150.1 | NP_001167621.1 | 7 | 1257 | ||
NM_001174151.1 | NP_001167622.1 | 6 | 1186 | |||
NM_001321328.1 | NP_001308257.1 | 8 | 1379 | |||
NM_144996.3 | NP_659433.2 | 5 | 936 | |||
NM_182896.2 | NP_878899.1 | 7 | 1257 | |||
NR_033427.1 | 6 | 1012 | ||||
NR_135621.1 | 6 | 1003 | ||||
XM_006713532.3 | XP_006713595.1 | 8 | 1376 | |||
XM_011512532.2 | XP_011510834.1 | 8 | 1285 | |||
XM_011512533.2 | XP_011510835.1 | 9 | 1093 | |||
XM_011512534.2 | XP_011510836.1 | 9 | 1411 | |||
XM_011512535.2 | XP_011510837.1 | 8 | 1270 | |||
XM_017005853.1 | XP_016861342.1 | 8 | 1287 | |||
GenBank mRNA | AK296994.1 | - | 7 | 1235 | ||
AK299284.1 | - | 6 | 996 | |||
AL713789.1 | - | 6 | 1129 | |||
BC015439.1 | - | 5 | 896 | |||
BC094725.1 | - | 5 | 931 | 4818884 | ||
BC104035.1 | - | 7 | 1247 | 40026398 | ||
BC104036.1 | - | 7 | 1247 | 40026399 | ||
CR936753.1 | - | 5 | 934 | |||
CR936756.1 | - | 6 | 891 | |||
CR936763.1 | - | 8 | 1331 |