Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105580.2 | NP_001099050.1 | 3 | 297 | ||
NR_047685.1 | 3 | 297 | ||||
GenBank mRNA | CR749263.1 | - | 3 | 306 |