Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105571.2 | NP_001099041.1 | 6 | 814 | ||
NM_001220493.1 | NP_001207422.1 | 6 | 817 | |||
GenBank mRNA | AI091419.1 | - | 1 | 184 | ||
AI168267.1 | - | 1 | 194 | |||
BC147024.1 | - | 6 | 814 | 9056952 | ||
BC147025.1 | - | 6 | 814 | 9056953 | ||
BC171909.1 | - | 6 | 817 | 9054563 |