Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181716.2 | NP_859067.2 | 2 | 548 | ||
GenBank mRNA | AF514992.1 | - | 2 | 484 | ||
BC052604.1 | - | 2 | 558 | 5934088 | ||
BC137486.1 | - | 2 | 580 | 9021486 | ||
BC137487.1 | - | 2 | 580 | 9021487 |
Gene Symbol: PIGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025349.1 | XP_016880838.1 | 9 | 4196 | ||
XM_017025350.1 | XP_016880839.1 | 9 | 4191 | |||
XM_017025352.1 | XP_016880841.1 | 10 | 3364 | |||
XM_017025353.1 | XP_016880842.1 | 9 | 3216 | |||
XM_017025354.1 | XP_016880843.1 | 9 | 3332 | |||
XM_017025355.1 | XP_016880844.1 | 8 | 3184 | |||
XM_017025356.1 | XP_016880845.1 | 9 | 4421 |