Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144606.5 | NP_653207.1 | 6, 7 | 1129 | ||
NM_144997.5 | NP_659434.2 | 6, 7 | 1129 | |||
XM_011523714.2 | XP_011522016.1 | 7, 8 | 1180 | |||
XM_011523716.1 | XP_011522018.1 | 5, 6 | 1558 | |||
XM_011523718.2 | XP_011522020.1 | 6, 7 | 1064 | |||
XM_011523719.2 | XP_011522021.1 | 7, 8 | 1180 | |||
XM_011523721.2 | XP_011522023.1 | 6, 7 | 929 | |||
XM_017024304.1 | XP_016879793.1 | 8, 9 | 1349 | |||
XM_017024305.1 | XP_016879794.1 | 8, 9 | 1364 | |||
XM_017024306.1 | XP_016879795.1 | 7, 8 | 1353 | |||
XM_017024307.1 | XP_016879796.1 | 7, 8 | 1315 | |||
XM_017024308.1 | XP_016879797.1 | 8, 9 | 1491 | |||
GenBank mRNA | AF517523.1 | - | 6, 7 | 1080 | ||
BC015687.2 | - | 6, 7 | 1080 | 4684974 | ||
KU178888.1 | - | 3, 4 | 625 |