Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178836.3 | NP_849158.2 | 2 | 751 | ||
XM_017024310.1 | XP_016879799.1 | 2 | 747 | |||
GenBank mRNA | AK090899.1 | - | 2 | 728 | ||
AK295282.1 | - | 3 | 671 | |||
BC031263.1 | - | 2 | 737 | 5274952 |
Gene Symbol: FLCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309336.1 | - | 12 | 1521 |