Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142958.1 | NP_001136430.1 | 5 | 758 | ||
NM_152676.2 | NP_689889.1 | 5 | 786 | |||
XM_011525858.2 | XP_011524160.1 | 5 | 727 | |||
XM_011525859.1 | XP_011524161.1 | 5 | 727 | |||
XM_011525860.1 | XP_011524162.1 | 5 | 727 | |||
XM_017025603.1 | XP_016881092.1 | 6 | 768 | |||
XM_017025604.1 | XP_016881093.1 | 5 | 2796 | |||
GenBank mRNA | AK094215.1 | - | 5 | 698 | ||
BC029579.1 | - | 5 | 736 | 5268841 |