Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153365.2 | NP_699196.2 | 12 | 1344 | ||
XM_005248139.2 | XP_005248196.1 | 11 | 1158 | |||
XM_005248140.3 | XP_005248197.1 | 11 | 1108 | |||
XM_011513812.2 | XP_011512114.1 | 11 | 1207 | |||
XM_011513815.2 | XP_011512117.1 | 11 | 2443 | |||
XM_011513816.2 | XP_011512118.1 | 11 | 1152 | |||
XM_011513817.2 | XP_011512119.1 | 9 | 721 | |||
XM_017007876.1 | XP_016863365.1 | 13 | 1234 | |||
GenBank mRNA | AK074494.1 | - | 12 | 1277 | ||
AK296104.1 | - | 11 | 1146 | |||
BC066899.1 | - | 12 | 1282 | 4825556 |