Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000118.3 | NP_000109.1 | 6 | 1126 | ||
NM_001114753.2 | NP_001108225.1 | 6 | 1126 | |||
NM_001278138.1 | NP_001265067.1 | 6 | 866 | |||
GenBank mRNA | AK222669.1 | - | 6 | 1039 | ||
AK290389.1 | - | 6 | 1010 | |||
AK301171.1 | - | 6 | 866 | |||
AK302455.1 | - | 3 | 238 | |||
BC014271.2 | - | 6 | 860 | 3843174 | ||
BT006872.1 | - | 6 | 708 | |||
J05481.1 | - | 6 | 669 | |||
X72012.1 | - | 6 | 989 |