Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182539.3 | NP_872345.2 | 3 | 635 | ||
XM_005248872.1 | XP_005248929.1 | 3 | 1087 | |||
XM_005248873.2 | XP_005248930.1 | 3 | 1087 | |||
XM_005248874.2 | XP_005248931.1 | 3 | 346 | |||
XM_006715009.3 | XP_006715072.1 | 4 | 751 | |||
XM_011514336.2 | XP_011512638.1 | 3 | 908 | |||
XM_011514338.2 | XP_011512640.1 | 4 | 1141 | |||
XM_011514339.2 | XP_011512641.1 | 3 | 357 | |||
XM_011514340.1 | XP_011512642.1 | 3 | 1087 | |||
GenBank mRNA | AK301825.1 | - | 3 | 635 | ||
BC035022.1 | - | 3 | 607 | 4824970 |