Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001977.3 | NP_001968.3 | 4 | 1303 | ||
GenBank mRNA | AK313412.1 | - | 5 | 965 | ||
AM888273.1 | - | 4 | 1303 | |||
AM888274.1 | - | 4 | 1303 | |||
AM888275.1 | - | 4 | 1303 | |||
AM888276.1 | - | 4 | 1303 | |||
AM888277.1 | - | 4 | 1303 | |||
AM888278.1 | - | 4 | 1303 | |||
AM888279.1 | - | 4 | 1303 | |||
AM888280.1 | - | 4 | 1303 | |||
AM888281.1 | - | 4 | 1303 | |||
AM888282.1 | - | 4 | 1303 | |||
AM888283.1 | - | 4 | 1303 | |||
BC094770.1 | - | 5 | 1284 | 30528477 | ||
L12468.1 | - | 5 | 1207 | |||
L14721.1 | - | 5 | 1044 |