Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293212.1 | NP_001280141.1 | 3, 4 | 662 | ||
NM_001293213.1 | NP_001280142.1 | 3, 4 | 578 | |||
NM_001293214.1 | NP_001280143.1 | 2, 3 | 446 | |||
NM_001293215.1 | NP_001280144.1 | 3, 4 | 565 | |||
NM_001293216.1 | NP_001280145.1 | 3, 4 | 464 | |||
NM_178014.3 | NP_821133.1 | 3, 4 | 578 | |||
GenBank mRNA | AB062393.1 | - | 3, 4 | 404 | ||
AF070561.1 | - | 3, 4 | 399 | |||
AF070593.1 | - | 3, 4 | 399 | |||
AF070600.1 | - | 3, 4 | 416 | |||
AF141349.1 | - | 3, 4 | 416 | |||
AK098772.1 | - | 3, 4 | 423 | |||
AK297489.1 | - | 3, 4 | 332 | |||
AK298889.1 | - | 3, 4 | 468 | |||
AK302314.1 | - | 3, 4 | 531 | |||
AK309684.1 | - | 1, 2 | 20 | |||
AK316265.1 | - | 3, 4 | 335 | |||
AK316316.1 | - | 3, 4 | 460 | |||
BC001938.1 | - | 3, 4 | 403 | 2820585 | ||
BC002347.2 | - | 3, 4 | 408 | 2964461 | ||
BC005838.2 | - | 3, 4 | 403 | 2820585 | ||
BC007605.1 | - | 3, 4 | 408 | 3350604 | ||
BC013374.2 | - | 3, 4 | 399 | 3946253 | ||
BC019924.1 | - | 3, 4 | 396 | 4998571 | ||
BC020946.1 | - | 3, 4 | 437 | 4795692 | ||
BC021909.1 | - | 3, 4 | 398 | 3509417 | ||
BC070326.1 | - | 3, 4 | 414 | 6698679 | ||
BC103746.1 | - | 3, 4 | 421 | 6503977 | ||
BU557299.1 | - | 3, 4 | 578 | |||
DC353572.1 | - | 3, 4 | 566 | |||
DC389555.1 | - | 2, 3 | 290 | |||
DC405540.1 | - | 3, 4 | 375 | |||
V00599.1 | - | 2, 3 | 251 |