Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313994.1 | NP_001300923.1 | 4, 5 | 524 | ||
NM_145004.6 | NP_659441.3 | 5, 6 | 604 | |||
XM_006716298.2 | XP_006716361.1 | 5, 6 | 398 | |||
XM_006716299.1 | XP_006716362.1 | 5, 6 | 398 | |||
XM_011544433.1 | XP_011542735.1 | 5, 6 | 398 | |||
XM_011544434.1 | XP_011542736.1 | 5, 6 | 398 | |||
XM_017013183.1 | XP_016868672.1 | 5, 6 | 398 | |||
XM_017013184.1 | XP_016868673.1 | 5, 6 | 398 | |||
XM_017013185.1 | XP_016868674.1 | 4, 5 | 404 | |||
XM_017013186.1 | XP_016868675.1 | 5, 6 | 398 | |||
GenBank mRNA | AB209690.1 | - | 5, 6 | 385 | ||
AK122608.1 | - | 5, 6 | 398 | |||
AK300148.1 | - | 4, 5 | 482 | |||
AK313864.1 | - | 5, 6 | 425 | |||
AY172952.1 | - | 5, 6 | 526 | |||
AY358739.1 | - | 5, 6 | 484 | |||
BC026085.2 | - | 5, 6 | 388 | 4838404 | ||
BC026169.2 | - | 5, 6 | 415 | 4829390 | ||
BC028702.2 | - | 5, 6 | 398 | 4829385 | ||
BC030014.2 | - | 5, 6 | 391 | 4830659 | ||
BC030698.2 | - | 5, 6 | 406 | 4822988 | ||
BC034975.2 | - | 5, 6 | 399 | 4829806 |