Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287005.1 | NP_001273934.1 | 6 | 911 | ||
NM_001287006.1 | NP_001273935.1 | 4 | 665 | |||
NM_001287007.1 | NP_001273936.1 | 3 | 476 | |||
NM_001287008.1 | NP_001273937.1 | 4 | 608 | |||
NM_145006.3 | NP_659443.1 | 5 | 797 | |||
XM_011518358.1 | XP_011516660.1 | 8 | 1991 | |||
XM_011518359.1 | XP_011516661.1 | 7 | 1941 | |||
XM_011518361.2 | XP_011516663.1 | 8 | 1053 | |||
XM_011518362.1 | XP_011516664.1 | 6 | 1827 | |||
XM_017014448.1 | XP_016869937.1 | 6 | 1098 | |||
XM_017014449.1 | XP_016869938.1 | 5 | 979 | |||
XM_017014450.1 | XP_016869939.1 | 7 | 938 | |||
XM_017014451.1 | XP_016869940.1 | 5 | 1547 | |||
GenBank mRNA | AA490088.1 | - | 2 | 170 | ||
AK128289.1 | - | 3 | 2765 | |||
AL137623.1 | - | 347 | ||||
AY358190.1 | - | 6 | 839 | |||
BC014601.2 | - | 5 | 778 | 4821517 | ||
BC041834.1 | - | 3 | 2691 | 5268318 | ||
BM458621.1 | - | 3 | 354 | |||
DR007158.1 | - | 4 | 641 |