Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001430.4 | NP_001421.2 | 12 | 2321 | ||
XM_011532698.2 | XP_011531000.1 | 12 | 2079 | |||
GenBank mRNA | AK123845.1 | - | 1 | 784 | ||
BC051338.1 | - | 12 | 2126 | 6305604 | ||
U81984.1 | - | 12 | 1960 |