Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005232.4 | NP_005223.4 | 12 | 2139 | ||
XM_006715880.3 | XP_006715943.1 | 11 | 1767 | |||
GenBank mRNA | AK290351.1 | - | 12 | 2103 | ||
AK313263.1 | - | 12 | 2112 | |||
BC130291.1 | - | 12 | 2123 | 40146322 | ||
BC144156.1 | - | 11 | 2275 | |||
CN289901.1 | - | 5 | 500 | |||
M18391.1 | - | 12 | 2169 | |||
Z27409.1 | - | 8 | 1197 |