Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018941.3 | NP_061764.2 | 2 | 820 | ||
XM_005266021.3 | XP_005266078.1 | 2 | 764 | |||
XM_005266022.1 | XP_005266079.1 | 2 | 943 | |||
XM_005266023.1 | XP_005266080.1 | 2 | 661 | |||
XM_011534745.1 | XP_011533047.1 | 2 | 765 | |||
XM_011534746.2 | XP_011533048.1 | 2 | 662 | |||
XM_011534747.1 | XP_011533049.1 | 2 | 820 | |||
GenBank mRNA | AK312486.1 | - | 2 | 763 | ||
BC007725.2 | - | 2 | 674 | 4303103 | ||
BT007049.1 | - | 1 | 515 |