Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159969.1 | NP_001153441.1 | 4, 5 | 909 | ||
NM_001981.2 | NP_001972.1 | 16, 17 | 1778 | |||
XM_005270618.2 | XP_005270675.1 | 15, 16 | 1719 | |||
XM_017000615.1 | XP_016856104.1 | 16, 17 | 1920 | |||
XM_017000616.1 | XP_016856105.1 | 15, 16 | 1716 | |||
XM_017000617.1 | XP_016856106.1 | 15, 16 | 1605 | |||
XM_017000618.1 | XP_016856107.1 | 14, 15 | 1515 | |||
XM_017000619.1 | XP_016856108.1 | 17, 18 | 1802 | |||
XM_017000620.1 | XP_016856109.1 | 16, 17 | 1969 | |||
GenBank mRNA | AK090936.1 | - | 4, 5 | 635 | ||
AK294297.1 | - | 5, 6 | 758 | |||
AK313396.1 | - | 16, 17 | 1727 | |||
BX647676.1 | - | 4, 5 | 890 | |||
CD657119.1 | - | 3, 4 | 553 | |||
DQ367924.1 | - | 16, 17 | 1686 | |||
U07707.1 | - | 16, 17 | 1707 | |||
Z29064.1 | - | 16, 17 | 1778 |