Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001432.2 | NP_001423.1 | 2, 3 | 308 | ||
GenBank mRNA | AK314959.1 | - | 2, 3 | 308 | ||
BC136404.1 | - | 2, 3 | 265 | 9020391 | ||
BC136405.1 | - | 2, 3 | 265 | 9020392 | ||
CR541887.1 | - | 2, 3 | 142 | |||
D30783.1 | - | 2, 3 | 308 |