Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301012.1 | NP_001287941.1 | 18 | 2055 | ||
NM_001301013.1 | NP_001287942.1 | 20 | 2235 | |||
NM_004100.4 | NP_004091.3 | 20 | 2359 | |||
NM_172103.3 | NP_742101.2 | 19 | 2290 | |||
NM_172105.3 | NP_742103.1 | 20 | 2359 | |||
XM_005266851.4 | XP_005266908.1 | 20 | 2084 | |||
XM_005266853.4 | XP_005266910.1 | 19 | 1997 | |||
XM_017010374.1 | XP_016865863.1 | 18 | 2221 | |||
XM_017010375.1 | XP_016865864.1 | 18 | 2195 | |||
GenBank mRNA | AK299378.1 | - | 21 | 2149 | ||
AK301950.1 | - | 19 | 2295 | |||
BC014193.2 | - | 12 | 1216 | |||
BC041063.1 | - | 19 | 2229 | 5458911 | ||
KU178044.1 | - | 18 | 1832 | |||
KU178045.1 | - | 17 | 1739 | |||
Y17114.1 | - | 20 | 2342 |