Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000123.3 | NP_000114.2 | 15 | 3529 | ||
GenBank mRNA | AF462447.1 | - | 15 | 3486 | ||
AK294708.1 | - | 15 | 3458 | |||
BC031522.2 | - | 15 | 3515 | 3869893 | ||
BQ002437.1 | - | 1 | 570 | |||
BX647399.1 | - | 15 | 3358 | |||
D16305.1 | - | 15 | 3110 | |||
L20046.1 | - | 15 | 3151 | |||
LM994821.1 | - | 11 | 2599 | |||
X69978.1 | - | 15 | 3300 |
Gene Symbol: BIVM-ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204425.1 | NP_001191354.1 | 23 | 4587 | ||
GenBank mRNA | AB209312.1 | - | 23 | 4379 | ||
LM994822.1 | - | 23 | 4465 |