Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001436.3 | NP_001427.2 | 5, 6 | 663 | ||
XM_005258651.2 | XP_005258708.1 | 5, 6 | 641 | |||
XM_011526623.2 | XP_011524925.1 | 4, 5 | 444 | |||
GenBank mRNA | AB451384.1 | - | 4, 5 | 549 | ||
AK296950.1 | - | 5, 6 | 628 | |||
BC015218.2 | - | 3, 4 | 368 | |||
BC019260.1 | - | 5, 6 | 617 | 3504198 | ||
BC019609.1 | - | 3, 4 | 367 | |||
BT006830.1 | - | 4, 5 | 549 | |||
BT020144.1 | - | 4, 5 | 549 | |||
CR457069.1 | - | 4, 5 | 549 | |||
M59849.1 | - | 5, 6 | 577 | |||
X56597.1 | - | 5, 6 | 608 |