Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001606.4 | NP_001597.2 | 37 | 5866 | ||
NM_212533.2 | NP_997698.1 | 37 | 5858 | |||
XM_006716996.3 | XP_006717059.1 | 36 | 5800 | |||
GenBank mRNA | AB028985.2 | - | 23 | 3723 | ||
AB177854.1 | - | 24 | 3767 | |||
AF178941.1 | - | 36 | 5768 | |||
AF327657.1 | - | 36 | 5724 | |||
AK295489.1 | - | 3 | 334 | |||
BC008755.1 | - | 8 | 1013 | |||
BC144588.1 | - | 36 | 5848 | |||
DB168116.1 | - | 3 | 326 |