Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163147.1 | NP_001156619.1 | 2, 3 | 591 | ||
NM_001163148.1 | NP_001156620.1 | 3, 4 | 384 | |||
NM_001163149.1 | NP_001156621.1 | 2, 3 | 255 | |||
NM_004956.4 | NP_004947.2 | 3, 4 | 788 | |||
NR_120445.1 | 3, 4 | 483 | ||||
XM_005249635.4 | XP_005249692.1 | 2, 3 | 301 | |||
XM_005249636.4 | XP_005249693.1 | 2, 3 | 224 | |||
XM_011515168.2 | XP_011513470.1 | 2, 3 | 234 | |||
GenBank mRNA | AB209202.1 | - | 2, 3 | 238 | ||
AK294755.1 | - | 2, 3 | 223 | |||
AK312863.1 | - | 3, 4 | 470 | |||
BC098403.1 | - | 3, 4 | 462 | 30345383 | ||
BC106762.1 | - | 2, 3 | 281 | 40025690 | ||
BC106763.1 | - | 2, 3 | 280 | 40025693 | ||
CN396908.1 | - | 3, 4 | 415 | |||
CN396911.1 | - | 2, 3 | 220 | |||
DB093597.1 | - | 2, 3 | 233 | |||
DB099964.1 | - | 3, 4 | 344 | |||
U17163.1 | - | 1, 2 | 49 | |||
X87175.1 | - | 2, 3 | 267 |