Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243280.1 | NP_001230209.1 | 4 | 1102 | ||
NM_001243281.1 | NP_001230210.1 | 4 | 1102 | |||
NM_001627.3 | NP_001618.2 | 4 | 1102 | |||
GenBank mRNA | AK054632.1 | - | 4 | 1055 | ||
AK300362.1 | - | 4 | 478 | |||
AK301804.1 | - | 5 | 535 | |||
AK316113.1 | - | 4 | 906 | |||
AL833702.1 | - | 4 | 903 | |||
BC137096.1 | - | 4 | 530 | 9021093 | ||
BC137097.1 | - | 4 | 530 | 9021094 | ||
DQ486139.1 | - | 4 | 468 | |||
DQ486140.1 | - | 4 | 468 | |||
L38608.1 | - | 4 | 469 | |||
Y10183.1 | - | 4 | 403 |