Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001993.4 | NP_001984.1 | 5 | 943 | ||
GenBank mRNA | AK312638.1 | - | 5 | 839 | ||
AY940730.1 | - | 5 | 723 | |||
BC011029.1 | - | 5 | 890 | 4073183 | ||
BT019808.1 | - | 5 | 722 | |||
CR541792.1 | - | 5 | 722 | |||
J02931.1 | - | 5 | 833 | |||
M16553.1 | - | 5 | 797 | |||
M27436.1 | - | 5 | 821 | |||
X07680.1 | - | 1 | 80 |