Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001443.2 | NP_001434.1 | 1, 2 | 183 | ||
GenBank mRNA | BC022287.1 | - | 1, 2 | 168 | ||
BC032801.1 | - | 1, 2 | 94 | 5190267 | ||
BP263155.1 | - | 1, 2 | 27 | |||
CR542090.1 | - | 1, 2 | 69 | |||
M10050.1 | - | 1, 2 | 111 | |||
M10617.1 | - | 1, 2 | 84 |