Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTIF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166261.1 | NP_001159733.1 | 3 | 270 | ||
NM_001166262.1 | NP_001159734.1 | 3 | 340 | |||
NM_001166263.1 | NP_001159735.1 | 4 | 369 | |||
NM_152912.4 | NP_690876.3 | 3 | 308 | |||
XM_006719771.3 | XP_006719834.1 | 4 | 356 | |||
XM_006719772.3 | XP_006719835.1 | 3 | 587 | |||
XM_011534957.2 | XP_011533259.1 | 4 | 830 | |||
XM_011534958.2 | XP_011533260.1 | 4 | 614 | |||
XM_011534959.2 | XP_011533261.1 | 4 | 779 | |||
XM_011534960.2 | XP_011533262.1 | 4 | 444 | |||
XM_011534961.2 | XP_011533263.1 | 4 | 1032 | |||
XM_011534962.2 | XP_011533264.1 | 4 | 609 | |||
XM_011534963.2 | XP_011533265.1 | 4 | 1197 | |||
XM_017020417.1 | XP_016875906.1 | 3 | 214 | |||
GenBank mRNA | AF410851.1 | - | 2 | 175 | ||
AK094420.1 | - | 1 | 1369 | |||
BC039599.1 | - | 3 | 261 | 3855096 | ||
BC046166.1 | - | 3 | 280 | 3460180 | ||
BG617536.1 | - | 3 | 309 | |||
BI460170.1 | - | 4 | 363 | |||
BM548061.1 | - | 2 | 233 | |||
CN272769.1 | - | 4 | 293 | |||
DB456678.1 | - | 3 | 342 | |||
DB470026.1 | - | 3 | 270 |