Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR5AR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004730.1 | NP_001004730.1 | 1 | 814 | ||
GenBank mRNA | BC140745.1 | - | 1 | 838 | 9021613 |