Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004104.4 | NP_004095.4 | 42 | 7320 | ||
XM_011523538.2 | XP_011521840.1 | 42 | 7388 | |||
GenBank mRNA | AB209988.1 | - | 42 | 7315 | ||
AY451392.1 | - | 41 | 7203 | |||
BC007267.1 | - | 16 | 2462 | |||
BC007909.1 | - | 11 | 1735 | |||
BC014631.2 | - | 18 | 3309 | |||
BC014634.2 | - | 5 | 731 | |||
BC063242.1 | - | 42 | 7320 | 6172538 | ||
S80437.1 | - | 7 | 1074 | |||
U26644.1 | - | 41 | 7185 | |||
U29344.1 | - | 42 | 7320 |