Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178505.7 | NP_848600.2 | 3 | 706 | ||
NR_134507.1 | 3 | 706 | ||||
NR_134508.1 | 3 | 706 | ||||
XM_011539450.2 | XP_011537752.1 | 3 | 494 | |||
XM_011539451.1 | XP_011537753.1 | 3 | 494 | |||
XM_017015829.1 | XP_016871318.1 | 3 | 494 | |||
GenBank mRNA | AK125611.1 | - | 3 | 494 | ||
BC042872.1 | - | 3 | 423 | 5164050 | ||
CR749606.1 | - | 3 | 706 |