Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282745.1 | NP_001269674.1 | 19 | 2666 | ||
NM_001282747.1 | NP_001269676.1 | 16 | 2198 | |||
NM_173081.4 | NP_775104.2 | 19 | 2687 | |||
XM_005252380.2 | XP_005252437.1 | 18 | 2475 | |||
XM_005252381.2 | XP_005252438.1 | 18 | 2508 | |||
XM_011519351.2 | XP_011517653.1 | 10 | 1517 | |||
XM_017015831.1 | XP_016871320.1 | 18 | 2454 | |||
GenBank mRNA | AK098711.1 | - | 9 | 1227 | ||
AK302104.1 | - | 16 | 2145 | |||
AK302985.1 | - | 19 | 2653 | |||
BC039312.1 | - | 19 | 2680 | 5296302 |