Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR8G5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005198.1 | NP_001005198.1 | 1 | 571 | ||
XM_011542662.2 | XP_011540964.1 | 4 | 1125 | |||
XM_011542663.2 | XP_011540965.1 | 4 | 895 | |||
XM_011542664.2 | XP_011540966.1 | 3 | 764 | |||
XM_011542665.2 | XP_011540967.1 | 3 | 1045 | |||
XM_011542666.2 | XP_011540968.1 | 3 | 815 | |||
XM_011542667.2 | XP_011540969.1 | 2 | 684 | |||
GenBank mRNA | BC136820.1 | - | 1 | 594 | 9020811 | |
BC136822.1 | - | 1 | 594 | 9020813 |