Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEPACAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152722.4 | NP_689935.2 | 2 | 765 | ||
XM_005271449.2 | XP_005271506.1 | 2 | 765 | |||
XM_017017361.1 | XP_016872850.1 | 2 | 765 | |||
GenBank mRNA | AK098396.1 | - | 2 | 403 | ||
AK122595.1 | - | 2 | 403 | |||
AY047587.1 | - | 2 | 403 | |||
BC104831.1 | - | 2 | 387 | 8143834 | ||
BC113562.1 | - | 2 | 387 | 8322614 |