Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002000.3 | NP_001991.1 | 4 | 645 | ||
NM_133269.3 | NP_579803.1 | 4 | 645 | |||
NM_133272.3 | NP_579806.1 | 3 | 609 | |||
NM_133274.3 | NP_579808.1 | 3 | 609 | |||
NM_133277.3 | NP_579811.1 | 2 | 318 | |||
NM_133278.3 | NP_579812.1 | 3 | 609 | |||
XM_011526625.2 | XP_011524927.1 | 4 | 569 | |||
XM_017026473.1 | XP_016881962.1 | 4 | 618 | |||
XM_017026474.1 | XP_016881963.1 | 3 | 401 | |||
GenBank mRNA | AK290922.1 | - | 4 | 503 | ||
BC027953.1 | - | 4 | 472 | 5211453 | ||
CV029531.1 | - | 4 | 448 | |||
D87853.1 | - | 4 | 448 | |||
D87855.1 | - | 3 | 412 | |||
D87857.1 | - | 3 | 412 | |||
D87859.1 | - | 5 | 526 | |||
D87861.1 | - | 2 | 121 | |||
D87862.1 | - | 3 | 412 | |||
DQ075334.1 | - | 4 | 448 | |||
DQ075335.1 | - | 4 | 448 | |||
DQ075336.1 | - | 4 | 448 | |||
DQ075337.1 | - | 4 | 448 | |||
DQ075338.1 | - | 4 | 448 | |||
DQ075339.1 | - | 4 | 448 | |||
U43774.1 | - | 4 | 453 | |||
U56236.1 | - | 4 | 458 | |||
U56237.1 | - | 3 | 422 | |||
X92558.1 | - | 3 | 412 | |||
X92559.1 | - | 3 | 412 | |||
X92560.1 | - | 4 | 448 |