Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000139.4 | NP_000130.1 | 3 | 342 | ||
XM_005273846.4 | XP_005273903.1 | 4 | 657 | |||
XM_011544850.2 | XP_011543152.1 | 4 | 275 | |||
XM_017017362.1 | XP_016872851.1 | 4 | 657 | |||
GenBank mRNA | AK304487.1 | - | 3 | 342 | ||
AK310142.1 | - | 3 | 342 | |||
BC074800.2 | - | 3 | 271 | 30915525 | ||
BC074843.2 | - | 3 | 271 | 30915222 | ||
D10583.1 | - | 3 | 290 | |||
DA972757.1 | - | 3 | 342 |