Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM171A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010924.1 | NP_001010924.1 | 5 | 635 | ||
XM_011519378.2 | XP_011517680.1 | 5 | 1295 | |||
XM_017015904.1 | XP_016871393.1 | 5 | 556 | |||
XM_017015905.1 | XP_016871394.1 | 4 | 329 | |||
GenBank mRNA | AL050367.1 | - | 5 | 635 | ||
AY683003.1 | - | 5 | 682 | |||
BC109302.1 | - | 5 | 635 | 40035211 | ||
BC109303.1 | - | 5 | 635 | 40035213 |