Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152727.5 | NP_689940.3 | 14 | 1505 | ||
GenBank mRNA | AF492484.1 | - | 14 | 1456 | ||
AK292269.1 | - | 13 | 1414 | |||
AK315394.1 | - | 14 | 1408 | |||
BC053658.1 | - | 15 | 1579 | 5750192 | ||
CR749617.1 | - | 13 | 1166 |