Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152730.5 | NP_689943.4 | 31 | 3588 | ||
NR_104452.1 | 35 | 4555 | ||||
XM_005266861.2 | XP_005266918.1 | 32 | 3711 | |||
XM_011535569.1 | XP_011533871.1 | 33 | 3774 | |||
XM_011535570.1 | XP_011533872.1 | 34 | 3822 | |||
XM_011535571.2 | XP_011533873.1 | 34 | 4475 | |||
XM_011535574.1 | XP_011533876.1 | 33 | 3770 | |||
XM_011535580.2 | XP_011533882.1 | 24 | 2625 | |||
XM_017010397.1 | XP_016865886.1 | 33 | 3759 | |||
XM_017010398.1 | XP_016865887.1 | 33 | 4407 | |||
XM_017010399.1 | XP_016865888.1 | 33 | 3699 | |||
XM_017010400.1 | XP_016865889.1 | 33 | 3696 | |||
XM_017010401.1 | XP_016865890.1 | 32 | 3636 | |||
XM_017010404.1 | XP_016865893.1 | 19 | 2045 | |||
GenBank mRNA | AK055461.1 | - | 15 | 1533 | ||
AK125385.1 | - | 21 | 2421 | |||
AK126259.1 | - | 11 | 1157 | |||
AK131446.1 | - | 35 | 4535 |
Gene Symbol: LOC105377975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA260573.1 | - | 474 |